VWR®, Protection covers, chemical resistant Maximum
VWRU414004-461
- Colour:Yellow
- L×W:1828×1524 mm
- Pk:25
These chemical-resistant covers provide the highest level of protection available within the VWR® Protection line.
- Superior chemical repellence as well as liquid and particle holdout
- Double-coated fabric protects against hazardous liquids and toxic chemicals
- Maximum durability and tear strength
Engineered to optimise barrier protection and prevent penetration by an extensive list of chemicals.
: Tested and manufactured in an ISO certified facility under stringent process controls. Lot-controlled and validated through independent laboratory testing.