Order Entry
Japan
ContactUsLinkComponent
VWR®Advanced, Block heaters
VWR®Advanced, Block heaters
Catalog # VWRU75838-282
Supplier:  VWR Collection
undefined
VWR®Advanced, Block heaters
Catalog # VWRU75838-282
Supplier:  VWR Collection

Specifications

  • No. of blocks:
    2
  • Weight:
    3,9 kg
  • Description:
    Advanced dry block heater, with capacity of 2 blocks, 120V
  • Seq:
    002
  • Electrical:
    120 V, 50/60 Hz, 400 W
  • Temperature uniformity:
    ±0,1 °C (across similar blocks at 37 °C)
  • Dimensions:
    196W×348L×117H mm
  • Pk:
    1

Specifications

About this item

Designed for applications that require repeatable results and superior temperature stability.

  • Compact design with small footprint
  • Exceptional temperature uniformity and stability
  • LED display for temperature and time
  • Heats to 120 °C (245 °F)
  • Accommodates interchangeable, modular heating blocks
  • Optional temperature probe kit

Multi-purpose units are ideal for incubation and activation of cultures, enzyme reactions, immunoassays, melting/boiling points, and a variety of other laboratory procedures.

Each model accepts separate interchangeable modular blocks, which accommodate various sample enclosures such as vials, microplates, PCR strips, and a wide variety of tubes. Each block contains a thermometer well and features close tube-and-block contact for maximum heat retention.

Easy to use front-mounted controls with independent LED displays show actual and set point temperatures. Microprocessor PID temperature control offers a stability of 0,1 °C, while independent controls allow users to adjust temperature settings in ±0,1 °C increments. A timer will display elapsed time or when programmed to user-defined limit, will count down to zero and shut off unit. Audible alarm will sound when time reaches zero. Temperature calibration mode allows unit to be calibrated to an external temperature device.

Optional external temperature probe monitors actual block or sample temperature and display that temperature on the control panel. The PT100 RTD probe is designed to fit perfectly into the thermometer well of each modular block.

Integral support rod holder with locking knob accepts optional external temperature probe kit.

Certifications: Available with a NIST Traceable, ISO/IEC 17025 accredited calibration certificate from Troemner. TUV listed RoHS compliant.
Ordering information: Unit must be ordered with heating block(s). Unit includes a 2340 mm detachable, three-wire cord and plug.
Caution: To avoid possible electrical hazard, do not fill well or block opening with water or other fluids. Unit is designed as a dry bath/incubator only.