Order Entry
Japan
ContactUsLinkComponent
VWR®, Beakers, Low Form, Heavy Duty
VWR®, Beakers, Low Form, Heavy Duty
Catalog # VWRU10536-520
Supplier:  VWR Collection
Supplier #: 10536-520
undefined
VWR®, Beakers, Low Form, Heavy Duty
Catalog # VWRU10536-520
Supplier:  VWR Collection
Supplier #: 10536-520
Log in for availability
Select an Option
500 ml
Select an Option
500 - 3500 ml
Select an Option
4000 ml
Select an Option
06
Select an Option
1
About this item
VWR® low form, heavy duty beakers are mechanically stronger than standard beakers and are designed for longer life with increased user safety.
  • With reinforced rim and spout
  • Borosilicate glass fabrication
  • Thick wall and heavy base
Durable matte finish marking area and graduated scale printed in white colour.
Certifications: Manufactured in accordance to ASTM E960 Type-2 specification.
Frequently Bought Together