About this item
Provide reliable, durable, and comfortable hand protection during demanding cleanroom applications. Textured fingers enhance grip on components.
- Quality tested according to ASTM, FTMS, and IEST Standards
- Provide comfort, protection, and tactile sensitivity
- Yield exceptionally low particle counts
- Highly resistant to permeation
Recommended for ISO Class 4 (FED-STD-209E Class 10/M2.5) or higher cleanrooms
Manufactured from a synthetic copolymer of acrylonitrile, butadiene, and methylacrylic acid (components that comply with FDA CFR 21). High tensile strength and strong permeation resistance against a wide variety of solvents, acids, and bases. Non-staining, non-smudging gloves reduce the risk of contamination to parts or components being handled by the user, even during prolonged wear.
Static-dissipative to inhibit harmful static discharge. Silicone-free material minimizes ionic contaminants and nonvolatile residue (NVR) transfer. Contain no natural rubber latex. AQL level of 1,5 for pinholes. For single use.