Piperidine ≥99% (by GC), J.T.Baker®
2895-98
: Avantor
:
- Pack type:Steel drum, non-removable head
- Pk:52 L
- Seq:5382
- Assay (C₅H₁₁N) (by GC):≥99 %
- Water (H₂O):≤0.2 %
:
:85.15 g/mol
:0.86 g/cm³
:16.00 °C
:2401
:8,I
:GHS02
:GHS05
:GHS06
:110-89-4
:C₅H₁₁N
:MFCD00005979