


Zinc ≥99.8% (by EDTA titration), granular, BAKER ANALYZED® ACS (30 Mesh), J.T.Baker®
Catalog # 4248-01
Supplier: Avantor
CAS Number:
Specifications
- Pack type:Poly bottle, white, blue T/E cap
- Pk:500 g
- Seq:3645
Specifications
Specification Test Results
- Assay (Zn) (by EDTA titrn):≥99.8 %
- Iron (Fe):≤0.01 %
- Lead (Pb):≤0.01 %
- Suitability for Arsenic Determination:Passes Test
- Trace Impurities - Arsenic (As):≤0.100 ppm
- Mesh - on U.S. No. 30 Sieve:≤5 %
- Mesh - Thru U.S. No. 50 Sieve:≤30 %
- Meets Reagent Specifications for testing USP/NF monographs:≤30%
Specification Test Results
About this item
:
:65.39 g/mol
Merck Index:12,10255
Boiling Pt:908 °C (1013 hPa)
Melting Pt:420 °C
Density:7.14 g/cm³ (20 °C)
:Management of Change (MOC) category = N
EINECS:231-175-3
:GHS07
:GHS08
:GHS09
Storage Temperature:Ambient
CAS Number:7440-66-6
Formula:Zn
MDL Number:MFCD00011291