Order Entry
Japan
ContactUsLinkComponent
Triacetin (glycerol triacetate) ≥99% (by GC), J.T.Baker®
Triacetin (glycerol triacetate) ≥99% (by GC), J.T.Baker®
Catalog # W307-07
Supplier:  Avantor
CAS Number:  
undefined
Triacetin (glycerol triacetate) ≥99% (by GC), J.T.Baker®
Catalog # W307-07
Supplier:  Avantor
CAS Number:  

Specifications

  • Pack type:
    Glass bottle, amber, narrow mouth in single shipper
  • Pk:
    500 g
  • Seq:
    3645

Specifications

Specification Test Results

  • Assay (C₉H₁₄O₆) (by GC):
    ≥ 99 %
  • Appearance:
    Passes Test

Specification Test Results

About this item

 :
 :218.21 g/mol
Merck Index:13,09664
Boiling Pt:258 °C (1013 hPa)
Melting Pt:4 °C
Density:1.1596 g/cm³ (20 °C)
Flash Pt:138 °C (closed cup)
Storage Temperature:Ambient
EINECS:203-051-9
CAS Number:102-76-1
Formula:C₉H₁₄O₆
MDL Number:MFCD00008716