
Rhodamine B (C.I. 45170), J.T.Baker®
U872-03
: Avantor
:
- Pack type:Glass bottle, amber, wide mouth, black T/E cap
- Pk:25 g
- Seq:3308
- Sensitivity for Antimony:Passes Test
:
:479.02 g/mol
:13,08266
:199…201 °C
:1.31 g/cm³ (20 °C)
:201-383-9
:GHS07
:Ambient
:81-88-9
:C₂₈H₃₁ClN₂O₃
:MFCD00011931