

Isopropyl acetate ≥99.0% (by GC), BAKER ANALYZED®, J.T.Baker®
Supplier: Avantor
Certificates
About this item
CAS Number:108-21-4
:
Formula:C₅H₁₀O₂
MDL Number:MFCD00008877
:102.13 g/mol
Merck Index:13,05225
Boiling Pt:85 °C (1013 hPa)
Melting Pt:–62 °C
Density:0,8732 g/cm³ (20 °C)
EINECS:203-561-1
UN:1220
ADR:3,II
:Management of Change (MOC) category = N
Flash Pt:4 °C
:GHS02
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:Safer Human and Environmental Health
- Environmental certification:Chem21 Solvent Selection Guide recommended by default and after discussion
Specifications
Specification Test Results
- Assay (by GC):≥99.0 %
- Density (g/mL) at 25°C:0.86 - 0.87
- Residue after Evaporation:≤0.0050 %
- Acidity (as CH₃COOH):≤0.01 %
- Water (H₂O):≤0.1 %
Specification Test Results
Frequently Bought Together