


1,2-Dimethoxyethane ≥99.0% (by GC, corrected for water content), BAKER ANALYZED®, J.T.Baker®
Catalog # J331-07
Supplier: Avantor
CAS Number:
Specifications
- Pack type:Glass bottle, amber, narrow mouth
- Pk:500 mL
- Seq:2725
Specifications
Specification Test Results
- Assay (C₄H₁₀O₂) (by GC, corrected for water):≥99.0 %
- Acidity (as CH₃COOH):≤0.015 %
- Density (g/mL) at 20°C:0.865 - 0.871
- Identification (by IR):Passes Test
- Peroxide (as H₂O₂):≤25 ppm
- Refractive Index, n²ºD:1.379 - 1.381
- Residue after Evaporation:≤0.0020 %
- Water (H₂O)(by Karl Fischer titrn):≤0.2 %
- Boiling Range (1-95 mL)Lower:83 - 86 °C
- Boiling Range (1-95 mL)Upper:83 - 86 °C
- Meets Reagent Specifications for testing USP/NF monographs:83 - 86 °C
Specification Test Results
About this item
:
:90.12 g/mol
UN:2252
ADR:3,II
:GHS02
:GHS07
:GHS08
CAS Number:110-71-4
Formula:C₄H₁₀O₂
MDL Number:MFCD00008502