


Triethylamine ≥98% (by GC), J.T.Baker®
Catalog # W639-09
Supplier: Avantor
CAS Number:
Specifications
- Pack type:Glass bottle, amber, narrow mouth
- Pk:4 L
- Seq:5162
Specifications
Specification Test Results
- Assay ((C₂H₅)₃N) (by GC):≥98 %
Specification Test Results
About this item
:
:101.19 g/mol
Density:0.73 g/cm³
Flash Pt:–11.00 °C
UN:1296
ADR:3,II
:GHS02
:GHS05
:GHS06
CAS Number:121-44-8
Formula:(C₂H₅)₃N
MDL Number:MFCD00009051