Order Entry
Japan
ContactUsLinkComponent
J.T.Baker® BAKER INSTRA-ANALYZED®, Silver, 1000 µg/mL, AA Standard
J.T.Baker® BAKER INSTRA-ANALYZED®, Silver, 1000 µg/mL, AA Standard
Catalog # 6467-04
Supplier:  Avantor
CAS Number:  
undefined
J.T.Baker® BAKER INSTRA-ANALYZED®, Silver, 1000 µg/mL, AA Standard
Catalog # 6467-04
Supplier:  Avantor
CAS Number:  

Specifications

  • Concentration:
    1000 mg/L
  • Matrix:
    5% HNO₃
  • Pk:
    150 mL
  • Element:
    Silver
  • Symbol:
    Au

Specifications

About this item

Silver standard is prepared by the dissolution of silver metal of 99.99% spectral purity. The metal content of the solution is confirmed by titration with Sodium Chloride. The standardization is traceable to NIST SRM 999.

  • Standardised with NIST standard chemicals

The final concentration of each lot is determined by titrimetry or gravimetry.