Order Entry
Japan
ContactUsLinkComponent
J.T.Baker® BAKER INSTRA-ANALYZED®, Cobalt, AA Standard
J.T.Baker® BAKER INSTRA-ANALYZED®, Cobalt, AA Standard
Catalog # 6450-04
Supplier:  Avantor
CAS Number:  
undefined
J.T.Baker® BAKER INSTRA-ANALYZED®, Cobalt, AA Standard
Catalog # 6450-04
Supplier:  Avantor
CAS Number:  

Specifications

  • Concentration:
    1000 mg/L
  • Matrix:
    5% HNO₃
  • Pack type:
    Poly bottle for liquids, narrow mouth
  • Pk:
    150 mL
  • Element:
    Cobalt
  • Symbol:
    Co

Specifications

About this item

The standard is prepared by the careful dilution of the metal or its salt, usually 99,99% pure as verified by arc emission spectrometry.

  • Standardised with NIST standard chemicals
  • Maximum shelf life and stability

The final concentration of each lot is determined by titrimetry or gravimetry.