Order Entry
Japan
ContactUsLinkComponent
Sodium benzoate dust-free 99.0-101.0% (by anhydrous basis) NF, FCC, Macron Fine Chemicals™
Irritant
Sodium benzoate dust-free 99.0-101.0% (by anhydrous basis) NF, FCC, Macron Fine Chemicals™
Catalog # MACR1454-26
CAS Number:  
Irritant
Sodium benzoate dust-free 99.0-101.0% (by anhydrous basis) NF, FCC, Macron Fine Chemicals™
Catalog # MACR1454-26
CAS Number:  

Specifications

  • Pack type:
    Fiber drum, Leverpack®
  • Pk:
    45 kg
  • Seq:
    4388

Specifications

Specification Test Results

  • Food GMP Manufactured Product:
    99.0 - 101.0%
  • Meets F.C.C. Requirements:
    Passes Test
  • GMP Manufactured Product:
    Passes Test
  • Meets N.F. Requirements:
    Passes Test
  • CAUTION: For Manufacturing, processing or repackaging:
    ≤0.04%
  • Bulk Food Chemical:
    ≤1.5%
  • Bulk Pharmaceutical Chemical:
    99.0 - 101.0%
  • NF - Assay (C₇H₅NaO₂) (anhydrous basis):
    99.0 - 101.0 %
  • NF - Identification A:
    Passes Test
  • NF - Identification B:
    Passes Test
  • NF - Identification C:
    Passes Test
  • NF - Alkalinity:
    Passes Test
  • NF - Water (H₂O):
    ≤1.5 %
  • FCC - Assay (C₇H₅NaO₂) (anhydrous basis):
    99.0 - 101.0 %
  • FCC - Arsenic (As):
    ≤3 ppm
  • FCC - Alkalinity (as NaOH):
    ≤0.04 %
  • FCC - Identification A:
    Passes Test
  • FCC - Identification B:
    Passes Test
  • FCC - Identification C:
    Passes Test
  • FCC - Lead (Pb):
    ≤2 mg/kg
  • FCC - Water (H₂O):
    ≤1.5 %
  • Solubility:
    Passes Test
  • Appearance (white, extruded cylinder):
    Passes Test

Specification Test Results

About this item

 :
 :144.11 g/mol
Density:1.44 g/cm³
 :GHS07
CAS Number:532-32-1
Formula:C₆H₅COONa
MDL Number:MFCD00012463