
Potassium hydroxide 1 N in aqueous solution, BAKER ANALYZED® ACS volumetric solution, J.T.Baker®
Supplier: Avantor
Certificates
About this item
CAS Number:1310-58-3
:
Formula:KOH
MDL Number:MFCD00003553
:56.11 g/mol
Density:1.05 g/cm³
Storage Temperature:Ambient
EINECS:215-181-3
UN:1814
ADR:8,II
:Management of Change (MOC) category = NI
:GHS05
Specification Test Results
- For Laboratory,Research,or Manufacturing Use:0.995 - 1.005
- Normality:0.995 - 1.005
- Chloride (Cl):≤10 ppm
- Nitrogen Compounds (as N):≤1 ppm
- ACS - Heavy Metals (as Pb):≤1 ppm
- Trace Impurities - Iron (Fe):≤0.5 ppm
- Appearance:Passes Test
Specification Test Results
Frequently Bought Together