Order Entry
Japan
ContactUsLinkComponent
Lead(II,IV) oxide, red, TECHNICAL
Health
Irritant
Environment
Lead(II,IV) oxide, red, TECHNICAL
Catalog # 26572.295
Supplier:  VWR Chemicals
CAS Number:  
undefined
Health
Irritant
Environment
Lead(II,IV) oxide, red, TECHNICAL
Catalog # 26572.295
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle for solids
  • Pk:
    1 kg
  • Seq:
    4024

Specifications

Specification Test Results

  • Assay (calculated as PbO2):
    Min. 30 %

Specification Test Results

About this item

 :
Merck Index:12,05449
Boiling Pt:2682 °C (1013 hPa)
Melting Pt:830 °C
Density:9.1 g/cm³ (20 °C)
EINECS:215-235-6
UN:1479
ADR:5.1,II
 :685.6 g/mol
 :GHS08
 :GHS07
 :GHS09
CAS Number:1314-41-6
Formula:Pb₃O₄
MDL Number:MFCD00078491