Calcium hydrogen orthophosphate, anhydrous 98.0-103.0%, powder USP, FCC, Macron Fine Chemicals™
:7757-93-9
:
:CaHPO₄
:MFCD00010909
:136.06 g/mol
:13,01697
:370 °C
:2,89 g/cm³ (20 °C)
:Ambient
:231-826-1
- Food GMP Manufactured Product:—
- Meets F.C.C. Requirements:—
- Meets U.S.P Requirements:—
- GMP Manufactured Product:—
- CAUTION: For Manufacturing, processing or repackaging:—
- Bulk Pharmaceutical Chemical:—
- Bulk Food Chemical:—
- USP - Assay (CaHPO₄):97.5 - 102.5 %
- USP - Loss on Ignition:6.6 - 8.7 %
- USP - Identification A:Passes Test
- USP - Identification B:Passes Test
- USP - Acid-Insoluble Substances:≤0.2 %
- USP - Carbonate (CO₃):Passes Test
- USP - Chloride (Cl):≤0.25 %
- USP - Fluoride (F):≤0.005 %
- USP - Sulfate (SO₄):≤0.5 %
- USP - Arsenic (As):≤3 ppm
- USP - Barium (Ba):Passes Test
- FCC - Assay (CaHPO₄):98.0 - 103.0 %
- FCC - Loss on Ignition:7.0 - 8.5 %
- FCC - Identification A:Passes Test
- FCC - Identification B:Passes Test
- FCC - Fluoride (F):≤0.005 %
- FCC - Arsenic (As):≤3 ppm
- FCC - Lead (Pb):≤2 ppm
- Appearance:Passes Test
- Solubility:Passes Test
Frequently Bought Together