Order Entry
Japan
ContactUsLinkComponent
 
Magnesium oxide heavy 98.0-100.5% Ph. Eur.
  83540.290
 :  
undefined
Magnesium oxide heavy 98.0-100.5% Ph. Eur.
  83540.290
 :  

 

  • Pack type:
    Plastic bottle for solids
  • Pk:
    1 kg
  • Seq:
    4024

 

 

  • Assay (calculated on ignited substance):
    98.0 - 100.5 %
  • Appearance:
    Conforms (see CoA/CoS)
  • Untapped bulk density:
    ≥ 0.25 g/ml
  • Identification (Mg):
    Passes test
  • Appearance of solution:
    Passes test
  • Soluble substances:
    ≤ 2.0 %
  • Substances insoluble in CH3COOH:
    ≤ 0.1 %
  • Cl (Chloride):
    ≤ 0.1 %
  • SO4 (Sulphate):
    ≤ 1.0 %
  • Ca (Calcium):
    ≤ 1.5 %
  • Fe (Iron):
    ≤ 0.07 %
  • Heavy metals (as Pb):
    ≤ 30 ppm
  • Loss on ignition (900°C):
    ≤ 8.0 %
  • Residual solvents:
    Passes test
  • As (Arsenic):
    ≤ 4 ppm
  • Cd (Cadmium):
    ≤ 1 ppm
  • Co (Cobalt):
    ≤ 1 ppm
  • Hg (Mercury):
    ≤ 2 ppm
  • Ni (Nickel):
    ≤ 3 ppm
  • Pb (Lead):
    ≤ 3 ppm
  • V (Vanadium):
    ≤ 1 ppm
  • Conforms to Ph.Eur.:
    Passes test

 

 

 :
 :40.3 g/mol
 :12,05713
 :3600 °C (1013 hPa)
 :2800 °C
 :3.58 g/cm³ (20 °C)
 :Ambient
 :215-171-9
 :1309-48-4
 :MgO
 :MFCD00011109