Order Entry
Japan
ContactUsLinkComponent
Bismuth(III) oxide ≥99.0% (by EDTA titration), BAKER ANALYZED® for electronic ceramics, J.T.Baker®
Health
Bismuth(III) oxide ≥99.0% (by EDTA titration), BAKER ANALYZED® for electronic ceramics, J.T.Baker®
Catalog # 1159-R
Supplier:  Avantor
CAS Number:  
undefined
Health
Bismuth(III) oxide ≥99.0% (by EDTA titration), BAKER ANALYZED® for electronic ceramics, J.T.Baker®
Catalog # 1159-R
Supplier:  Avantor
CAS Number:  

Specifications

  • Pk:
    100 lb
  • Seq:
    7167

Specifications

Specification Test Results

  • For Microelectronic Use:
    = 99.0 %
  • Assay (as Bi₂O₃) (by EDTA titrn):
    ≥99.0 %
  • Chloride (Cl):
    ≤0.001 %
  • Nitrate (NO₃):
    ≤0.03 %
  • Sulfur Compounds (as SO₄):
    ≤0.002 %
  • Aluminum (Al):
    ≤0.005 %
  • Calcium (Ca):
    ≤0.005 %
  • Copper (Cu):
    ≤0.005 %
  • Iron (Fe):
    ≤0.005 %
  • Lead (Pb):
    ≤0.005 %
  • Silicon (Si):
    ≤0.01 %
  • Sodium (Na):
    ≤0.01 %
  • Average Particle Diameter, µm (APD):
    ≤15
  • Particle Diameter - at 90%, For Information Only:
    For Information Only
  • Specific Surface Area, m²/g, For Information Only:
    For Information Only
  • Bulk Density (g/cc), For Information Only:
    For Information Only

Specification Test Results

About this item

 :
 :465.96 g/mol
Merck Index:14,07243
Melting Pt:258…262 °C
Density:1.3 g/cm³ (20 °C)
EINECS:201-004-7
REACH:01-2119498295-24
 :GHS08
Storage Temperature:Ambient
CAS Number:1304-76-3
Formula:BiO₃
MDL Number:MFCD00003462