Order Entry
Japan
ContactUsLinkComponent
Iso-Hexane mixture of isomers ≥97.0% (mixture of isomers), HiPerSolv CHROMANORM® for HPLC
Iso-Hexane mixture of isomers ≥97.0% (mixture of isomers), HiPerSolv CHROMANORM® for HPLC
Catalog # 83622.320
Supplier:  VWR Chemicals
CAS Number:  
undefined
Iso-Hexane mixture of isomers ≥97.0% (mixture of isomers), HiPerSolv CHROMANORM® for HPLC
Catalog # 83622.320
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    2,5 L
  • Seq:
    5096

Specifications

Specification Test Results

  • Appearance:
    Clear colorless liquid
  • Assay (Mix. of C₆H₁₄ isomers):
    Min. 97.00 %
  • Water (KF):
    Max. 0.0100 % w/w
  • Residue after evaporation:
    Max. 0.0005 % w/w
  • n-Hexane:
    Max. 3.0 %
  • Transmittance (205 nm):
    Min. 50.0 %
  • Transmittance (220 nm):
    Min. 80.0 %
  • Transmittance (230 nm):
    Min. 95.0 %
  • Transmittance (280 nm):
    Min. 98.0 %

Specification Test Results

About this item

CAS Number:107-83-5
 :
Formula:C₆H₁₄
 :86.18 g/mol
Boiling Pt:60 °C (1013 hPa)
Melting Pt:–154 °C
Density:0.653 g/cm³ (25 °C)
Flash Pt:–33 °C
Storage Temperature:Ambient
EINECS:295-570-2
ADR:3,II
 :Filtered through a 0.2 µm filter, packaged under nitrogen.
UN:1208
REACH:01-2119484651-34
 :GHS02
 :GHS08
 :GHS07
 :GHS09
CAS Number:64742-49-0