$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Japan
ContactUsLinkComponent
Media for Microbiology
Prepared Media for Microbiology
VWR®, Dehydrated culture media in containers
VWR®, Dehydrated culture media in containers
Catalog # 7401.04500
Supplier:
VWR Chemicals
CAS Number:
Expand image
VWR®, Dehydrated culture media in containers
Catalog # 7401.04500
Supplier:
VWR Chemicals
CAS Number:
Specifications
Pk:
5
Description:
Media container, BPW
Sterile water to be added:
4500 ml/container
Specifications
About this item
Product Details
Product Details