VWR®, Peaked Caps
Supplier: VWR Collection
Certificates
About this item
These disposable caps, with peak, are made from a non woven, spunbonded polypropylene (SPP) fabric, which protects against dirt, grime, and certain dry particulates, in non hazardous environments.
- Free from natural rubber latex
- Available in white and blue
Weight: 40 g/m² (top), 100 g/m² (peak)
Packaging: 100 pieces per bag, 10 bags per case.
Related Information
Food contact certificate F
Additional Product Details
TDS VWR® Peaked Cap
Datasheet
VWR® Non Regulated Disposables for Production
Product Literature
Frequently Bought Together