Order Entry
Japan
ContactUsLinkComponent
Potassium citrate monohydrate ≥99.0%, AnalaR NORMAPUR® analytical reagent
Potassium citrate monohydrate ≥99.0%, AnalaR NORMAPUR® analytical reagent
Catalog # 102004S
Supplier:  VWR Chemicals
CAS Number:  
undefined
Potassium citrate monohydrate ≥99.0%, AnalaR NORMAPUR® analytical reagent
Catalog # 102004S
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle for solids
  • Pk:
    500 g
  • Seq:
    3645

Specifications

Specification Test Results

  • Assay (on anhydrous substance):
    Min. 99.0 %
  • Solution in water:
    Passes test
  • pH (20°C; 5 %):
    8.0 - 9.5
  • Total N (Nitrogen):
    Max. 10 ppm
  • C2O4 (Oxalate):
    Max. 100 ppm
  • Cl (Chloride):
    50 ppm
  • SO4 (Sulphate):
    Max. 50 ppm
  • Cu (Copper):
    Max. 1 ppm
  • Fe (Iron):
    Max. 5 ppm
  • Na (Sodium):
    Max. 0.2 %
  • Pb (Lead):
    Max. 5 ppm

Specification Test Results

About this item

 :
Merck Index:13,07706
Melting Pt:230 °C
Density:1.98 g/cm³ (20 °C)
 :324.41 g/mol
EINECS:212-755-5
CAS Number:6100-05-6
Formula:HOC(COOK)(CH₂COOK)₂·H₂O
MDL Number:MFCD00150442