
L(+)-Arginine 98.5-101.5% USP, Multi-Compendial, J.T.Baker®
2066-06
: Avantor
:
- Pack type:Plastic bottle
- Pk:1 kg
- Seq:4024
- USP - Assay (C₆H₁₄N₄O₂) (dried basis):98.5 - 101.5 %
- USP - Identification:Passes Test
- USP - Specific Rotation [Å]²⁵ ^D (+):26.3 - 27.7 Degree
- USP - Loss on Drying at 105°C:≤ 0.5 %
- USP - Chloride (Cl):≤ 0.05 %
- USP - Residue on Ignition:≤ 0.3 %
- USP - Sulfate (SO₄):≤ 0.03 %
- USP - Iron (Fe):≤ 0.003 %
- USP - Related Compounds - Individual Impurities:≤ 0.5 %
- USP - Related Compounds - Total Impurities:≤ 2.0 %
- EP/BP - Assay (C₆H₁₄N₄O₂) (dried basis):98.5 - 101.0 %
- EP/BP - Identification A:Passes Test
- EP/BP - Identification C:Passes Test
- EP/BP - Appearance of Solution:Passes Test
- EP/BP - Specific Rotation [Å]²º^D (+):25.5 - 28.5 Degree
- EP/BP-Ninhydrin-Positive Substances-Each:≤ 0.2 %
- EP/BP-Ninhydrin-Positive Substances-Total Impurities:≤ 0.5 %
- EP/BP - Chloride (Cl):≤ 200 ppm
- EP/BP - Sulfate (SO₄):≤ 300 ppm
- EP/BP - Ammonium (NH₄):≤ 0.02 %
- EP/BP - Iron (Fe):≤ 10 ppm
- EP/BP - Loss on Drying:≤ 0.5 %
- EP/BP - Ash (sulfated):≤ 0.1 %
- JP - Identification:Passes Test
- JP - Optical rotation, 20° C (+):26.9 - 27.9 Degree
- JP - pH (1:10):10.5 - 12.0
- JP - Clarity and Color of Solution:Passes Test
- JP - Chloride (Cl):≤ 0.021 %
- JP - Sulfate (SO₄):≤ 0.028 %
- JP - Ammonium (NH₄):≤ 0.02 %
- JP - Heavy Metals (as Pb):≤ 10 ppm
- JP - Iron (Fe):≤ 10 ppm
- JP - Related Substances:Passes Test
- JP - Loss on Drying at 105°C:≤ 0.30 %
- JP - Residue after Ignition:≤ 0.1 %
- JP - Assay (C₆H₁₄N₄O₂)(dried basis):98.5 - 101.0 %
- Endotoxin Concentration, IU/mg, For Information Only:≤ 0.1 %
- No Class 1,2,3 or other solvents are used or produced in the:98.5 - 101.0 %
- manufacturing or purification of the product.:= 10 ppm
- No Class 1,2,3 or other solvents are used or produced in the manufacturing or purification of the product.:= 10 ppm
- Elemental Impurities (USP (232), EP 5.20) - Information on elemental:Passes Test
- impurities for this product is available on the associated Product:= 0.3 %
- Regulatory Data Sheet and elemental impurity profile report.:= 0.1 %
- Endotoxin Concentration, IU/mg:For Information Only
:
:174.2 g/mol
:13,00785
:368 °C (1013 hPa)
:238 °C
:1.3 g/cm³ (20 °C)
:Management of Change (MOC) category = R
:Ambient
:200-811-1
:GHS07
:74-79-3
:C₆H₁₄N₄O₂
:MFCD00002635