Order Entry
Japan
ContactUsLinkComponent
tetra-Chloroauric(III) acid trihydrate, AnalaR NORMAPUR® analytical reagent
Corrosive
Irritant
tetra-Chloroauric(III) acid trihydrate, AnalaR NORMAPUR® analytical reagent
Catalog # 1028.0001
Supplier:  VWR Chemicals
CAS Number:  
undefined
Corrosive
Irritant
tetra-Chloroauric(III) acid trihydrate, AnalaR NORMAPUR® analytical reagent
Catalog # 1028.0001
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

Specification Test Results

  • Assay (Au):
    Min. 48 %
  • Alkali metals:
    Max. 0.05 %

Specification Test Results

About this item

 :
 :393.83 g/mol
Melting Pt:30 °C
Density:3.9 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:240-948-4
UN:3260
ADR:8,III
 :GHS05
 :GHS07
CAS Number:16961-25-4
Formula:HAuCl₄·3H₂O
MDL Number:MFCD00149904