Order Entry
Japan
ContactUsLinkComponent
tri-Sodium citrate dihydrate 99.0-100.5% (by anhydrous basis), crystalline, GenAR® USP, Macron Fine Chemicals™
tri-Sodium citrate dihydrate 99.0-100.5% (by anhydrous basis), crystalline, GenAR® USP, Macron Fine Chemicals™
Catalog # MACR0634-45
CAS Number:  
undefined
tri-Sodium citrate dihydrate 99.0-100.5% (by anhydrous basis), crystalline, GenAR® USP, Macron Fine Chemicals™
Catalog # MACR0634-45
CAS Number:  

Specifications

  • Pack type:
    Fibre drum
  • Pk:
    125 kg
  • Seq:
    4568

Specifications

Specification Test Results

  • GMP Manufactured Product:
    99.0 - 100.5%
  • Meets E.P. Chemical Specifications:
    Passes Test
  • Meets U.S.P Requirements:
    Passes Test
  • Meets J.P. Chemical Specifications:
    Passes Test
  • CAUTION: For Manufacturing, processing or repackaging:
    Passes Test
  • Bulk Pharmaceutical Chemical:
    Passes Test
  • USP - Assay (C₆H₅Na₃O₇) (anhydrous basis):
    99.0 - 100.5 %
  • USP - Appearance (colorless or white to slightly yellow crystals):
    Passes Test
  • USP - Identification A:
    Passes Test
  • USP - Identification B:
    Passes Test
  • USP - Identification C:
    Passes Test
  • USP - Alkalinity:
    Passes Test
  • USP - Water (H₂O):
    10.0 - 13.0 %
  • USP - Tartrate:
    Passes Test
  • EP - Assay (anhydrous basis):
    99.0 - 101.0 %
  • EP - Appearance of Solution (clear and colorless):
    Passes Test
  • EP - Identification A:
    Passes Test
  • EP - Identification B:
    Passes Test
  • EP - Acidity or Alkalinity:
    Passes Test
  • EP - Readily Carbonizable Substances:
    Passes Test
  • EP - Chlorides (Cl):
    ≤50 ppm
  • EP - Oxalate (C₂O₄):
    ≤300 ppm
  • EP - Sulfate (SO₄):
    ≤150 ppm
  • EP - Water (H₂O):
    11.0 - 13.0 %
  • JP - Assay (anhydrous basis):
    99.0 - 101.0 %
  • JP - Description:
    Passes Test
  • JP - Identification A:
    Passes Test
  • JP - Identification B:
    Passes Test
  • JP - pH:
    7.5 - 8.5
  • JP - Clarity and Color of Solution:
    Passes Test
  • JP - Chloride (Cl):
    ≤0.015 %
  • JP - Sulfate (SO₄):
    ≤0.048 %
  • JP - Heavy Metals:
    ≤10 ppm
  • JP - Arsenic (As):
    ≤2 ppm
  • JP - Tartrate:
    Passes Test
  • JP - Oxalate (C₂O₄):
    Passes Test
  • JP - Readily Carbonizable Substances:
    Passes Test
  • JP - Loss on Drying at 180°C:
    10.0 - 13.0 %

Specification Test Results

About this item

 :
 :294.1 g/mol
Merck Index:13,08675
Melting Pt:150 °C
Density:1.70 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:200-675-3
CAS Number:6132-04-3
Formula:Na₃C₆H₅O₇·2H₂O
MDL Number:MFCD00150031