
Oxalic acid dihydrate 99.5-102.5%, AnalaR NORMAPUR® ACS, Reag. Ph. Eur. analytical reagent
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:6153-56-6
:
Formula:HO₂CCO₂H·2H₂O
MDL Number:MFCD00149102
:126.07 g/mol
Merck Index:13,06980
Boiling Pt:149…160 °C (1013 hPa)
Melting Pt:101 °C
Density:1.65 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:205-634-3
UN:3261
ADR:8,III
REACH:01-2119534576-33
:GHS07
Specification Test Results
- Assay:99.5 - 102.5 %
- IR Spectrum:Passes test
- Reaction with sulphuric acid:Passes test
- Heavy metals (as Pb):Max. 5 ppm
- Ignition residue (SO4):Max. 100 ppm
- Insolubility in water:Max. 50 ppm
- Total N (Nitrogen):Max. 10 ppm
- Cl (Chloride):Max. 5 ppm
- SO4 (Sulphate):Max. 50 ppm
- Ca (Calcium):Max. 10 ppm
- Cu (Copper):Max. 5 ppm
- Fe (Iron):Max. 2 ppm
- Mg (Magnesium):Max. 5 ppm
- Pb (Lead):Max. 5 ppm
- Conforms to ACS:Passes test
- Conforms to Reag. Ph.Eur.:Passes test
Specification Test Results
Related Information
Certificates
Search by Lot Number2,5L Plastic bottle for solids DIN 80 (ZPACK13252500) 750 ml Plastic bottle for solids DIN 60 (ZPACK13250750)
Packaging Information
spec_VWRC20562.234 spec_VWRC20562.260 spec_VWRC20562.260 spec_VWRC20562.260 spec_VWRC20562.291
Product specification
More
Frequently Bought Together