Order Entry
Japan
ContactUsLinkComponent
L(+)-Cysteine hydrochloride monohydrate 98.5-101.5% (dry basis) USP, Multi-Compendial, J.T.Baker®
L(+)-Cysteine hydrochloride monohydrate 98.5-101.5% (dry basis) USP, Multi-Compendial, J.T.Baker®
Catalog # 2071-06
Supplier:  Avantor
CAS Number:  
undefined
L(+)-Cysteine hydrochloride monohydrate 98.5-101.5% (dry basis) USP, Multi-Compendial, J.T.Baker®
Catalog # 2071-06
Supplier:  Avantor
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle for solids
  • Pk:
    1 kg
  • Seq:
    4024

Specifications

Specification Test Results

  • USP - Assay (C₃H₇NO₂SHCl) (dried basis):
    98.5 - 101.5 %
  • USP - Identification:
    Passes Test
  • USP - Specific Rotation α²⁵D (+):
    5.7 - 6.8 Degree
  • USP - Loss on Drying:
    8.0 - 12.0 %
  • USP - Residue on Ignition:
    ≤ 0.1 %
  • USP - Sulfate (SO₄):
    ≤ 0.03 %
  • USP - Iron (Fe):
    ≤ 0.003 %
  • USP - Related Compounds - Individual Impurities:
    ≤ 0.5 %
  • USP - Related Compounds - Total Impurities:
    ≤ 2.0 %
  • FCC - Assay (C₃H₇NO₂SHCl) (dried basis):
    98.0 - 101.5 %
  • EP/BP - Assay (C₃H₇NO₂SHCl) (dried basis):
    98.5 - 101.0 %
  • EP/BP - Identification A:
    Passes Test
  • EP/BP - Identification B:
    Passes Test
  • EP/BP - Identification E:
    Passes Test
  • EP/BP - Appearance of Solution:
    Passes Test
  • EP/BP - Specific Rotationα²ºD (+):
    5.5 - 7.0 Degree
  • EP/BP - Ninhydrin-Positive Substances - Impurity A:
    ≤ 0.5 %
  • EP/BP-Ninhydrin-Positive Substances-Each:
    ≤ 0.2 %
  • EP/BP-Ninhydrin-Positive Substances-Total Impurities:
    ≤ 1.0 %
  • EP/BP - Sulfate (SO₄):
    ≤ 300 ppm
  • EP/BP - Ammonium (NH₄):
    ≤ 0.02 %
  • EP/BP - Iron (Fe):
    ≤ 20 ppm
  • EP/BP - Loss on Drying:
    8.0 - 12.0 %
  • EP/BP - Ash (sulfated):
    ≤ 0.1 %
  • Endotoxin Concentration, IU/mg, For Information Only:
    = 1.0 %
  • No Class 1,2,3 or other solvents are used or produced in the manufacturing or purification of the product.:
    = 300 ppm
  • Metallic Residues: No metal catalysts or metal reagents, as defined by EMA Guideline EMEA/CHMP/SWP/4446/2000 , are used in the production of this material.:
    = 0.02 %
  • EP/BP - Heavy Metals (as Pb):
    = 10 ppm
  • Endotoxin Concentration, IU/mg:
    For Information Only

Specification Test Results

About this item

 :
 :175.64 g/mol
Merck Index:13,02810
Melting Pt:168…170 °C
Storage Temperature:Ambient
EINECS:200-157-7
CAS Number:7048-04-6
Formula:HSCH₂CH(NH₂)COOH·HCl·H₂O
MDL Number:MFCD00065606