Order Entry
Japan
ContactUsLinkComponent
Paraffin, liquid Ph. Eur.
Paraffin, liquid Ph. Eur.
Supplier:  VWR Chemicals
Certificates
undefined
Paraffin, liquid Ph. Eur.
Supplier:  VWR Chemicals
Certificates

Specification Test Results

  • Appearance:
    Clear oily liquid
  • Identification A:
    Passes test
  • Acidity or alkalinity:
    Passes test
  • Relative density:
    0.81 - 0.875
  • Viscosity:
    25 - 80 mPa.s
  • Polycyclic aromatic hydrocarbons:
    Passes test
  • Readily carbonisable substances:
    Passes test
  • Solid paraffins:
    Passes test
  • Residual solvents:
    Unlikely by manuf.process

Specification Test Results

Related Information

Frequently Bought Together