Order Entry
Japan
ContactUsLinkComponent
Paraffin, Solidification point: 52-54°C, pellets Ph. Eur.
Paraffin, Solidification point: 52-54°C, pellets Ph. Eur.
Supplier:  VWR Chemicals
Certificates
undefined
Paraffin, Solidification point: 52-54°C, pellets Ph. Eur.
Supplier:  VWR Chemicals
Certificates

Specification Test Results

  • Appearance:
    White mass
  • Identification A:
    Passes test
  • Melting point:
    50 - 61 °C
  • Acidity or alkalinity:
    Passes test
  • Polycyclic aromatic hydrocarbons:
    Passes test
  • SO4 (Sulphate):
    Max. 150 ppm
  • Residual solvents:
    Passes test

Specification Test Results

Frequently Bought Together