Order Entry
Japan
ContactUsLinkComponent
Mercury, BAKER ANALYZED® ACS, J.T.Baker®
Toxic
Health
Environment
Mercury, BAKER ANALYZED® ACS, J.T.Baker®
Catalog # 2564-01
Supplier:  Avantor
CAS Number:  
undefined
Toxic
Health
Environment
Mercury, BAKER ANALYZED® ACS, J.T.Baker®
Catalog # 2564-01
Supplier:  Avantor
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    454 g
  • Seq:
    3635

Specifications

Specification Test Results

  • For Laboratory,Research,or Manufacturing Use:
    Passes Test
  • Appearance:
    Passes Test
  • Nonvolatile Matter:
    ≤5 ppm

Specification Test Results

About this item

 :
 :200.59 g/mol
Merck Index:13,05925
Boiling Pt:356.5 °C (1013 hPa)
Melting Pt:–39 °C
Density:13.54 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-106-7
UN:2809
ADR:8,III
 :GHS06
 :GHS08
 :GHS09
CAS Number:7439-97-6
Formula:Hg
MDL Number:MFCD00011035