Order Entry
Japan
Orders LinkContactUsLinkComponent
VWR®, Screw Caps
VWR®, Screw Caps
Supplier:   VWR Collection
VWR®, Screw Caps
Supplier:  VWR Collection
Certificates
VWR®, Screw Caps
VWR®, Screw Caps
3 Options Available Below
Product Family Options
Product Information
  • Thread
    For bottles
    Availability
    Price
  • 18 mm
    50/100 ml
    Log in for availability
    Log in for availability
    1 * 10 items
    1 * 10 items
  • 25 mm
    250/500 ml
    Log in for availability
    Log in for availability
    1 * 10 items
    1 * 10 items
  • 28 mm
    1000/2000 ml
    Log in for availability
    Log in for availability
    1 * 10 items
    1 * 10 items