Order Entry
Japan
Orders LinkContactUsLinkComponent
VWR®, Swab Applicator
VWR®, Swab Applicator
Swab applicators, 152 mm
Catalog # VWRU470019-172
Supplier:   VWR Collection
VWR®, Swab Applicator
Catalog # VWRU470019-172
Supplier:  VWR Collection
VWR®, Swab Applicator
VWR®, Swab Applicator
Swab applicators, 152 mm
Price
$6.29
Log in for availability
1 * 10 items
Product Details & Documents
This sterile cotton-tipped applicator spreads a confluent lawn of bacteria without breaking the agar surface.
  • Easy sub-culturing and bacterial transfers
Packaging: Individual.
Caution: *The product(s) on this page are outside the scope of the EU Directive 2019/904
Specifications
  • Description:
    Swab applicators
  • Length:
    152 mm