VWR®, Prepared Slides for Microbiology
VWRU470176-204
- Description:Prepared slides for microbiology
- Pk:12
Prepared slides to investigate the morphological differences between bacteria and fungi.
- Penicillum (wm)
- Yeast, budding (wm)
- Capsules of bacteria (sm)
- Typical coccus (sm)
- Typical spirillum (sm)
- Bacteria types (sm)
- Bacillus in chains (sm)
- Typical mixed bacteria (sm)
- Spirochaetes (sm)
- Treponema pallidium (sm)
- Typical bacillus (sm)
- Staphylococcus epidermidis (sm)
Designed for applications in microbiology.
(wm) = whole mount ; (sm) = smear.
: Supplied in a durable box.