VWR®, Chemical resistant aprons
Catalog # VWRU89232-928
Supplier: VWR Collection
Specifications
- Garment fit:Long
- Garment application:Industrial
- Gender:Unisex
- Colour:Yellow
- Length:1450 mm
- Sleeve or leg length:Long
- Pk:50
Specifications
About this item
These aprons are designed to provide protection against chemical spray in hazardous waste remediation, environmental cleanup, petrochemical/oil refining, acid/caustic handling, biohazard cleanup, and pharmaceutical manufacturing applications.
- Extra long to provide additional coverage below the knee
- Elastic wrists, open back, and bound seams for maximum protection