VWR®, Centrifuge Bottles, PP/PC, with Sealing Cap
Supplier: VWR Collection
Certificates
About this item
These precision molded centrifuge bottles are manufactured from PC or PP.
- Autoclavable
- Extra thick walls for durability resistant to most chemicals
- Wide mouth design makes filling and decanting easier
- Sealing caps have a built-in gripper to facilitate removal from the rotor
For high-speed work, the easy-to-use Noryl® sealing caps with silicone O-rings enable leak-free processing at high centrifugal forces over long durations.
Bottles are qualified to the rated g-forces using Sorvall® and Fiberlite® high-speed rotors, tested at 20 ºC, with bottles filled to 80% of capacity.
Caution: Maximum RCF ratings are based on using the proper rotor and/or support cushions. Without the proper support, these bottles may distort and/or fail.
Frequently Bought Together