VWR® Cell Chambers
Supplier: VWR Collection
About this item
VWR® cell chambers, made from medical grade, general purpose polystyrene (GPPS) and with vented cap, are ideal for large-scale cell culture.
- Available in 1-, 2-, 5- and 10-tray versions for easy scale-up
- Surface treated for optimal cell attachment and growth
- Hydrophobic membrane (pore size of 0,22 µm) avoids potential contamination during gas exchange
- Does not absorb cytokines and growth factors, and cell harvesting by conventional trypsin digestion method is possible
- Sterilised by irradiation
- Free from DNase and RNase, non pyrogenic
9 Options Available Below
Product Details & Documents
VWR® cell chambers, made from medical grade, general purpose polystyrene (GPPS) and with vented cap, are ideal for large-scale cell culture.
- Available in 1-, 2-, 5- and 10-tray versions for easy scale-up
- Surface treated for optimal cell attachment and growth
- Hydrophobic membrane (pore size of 0,22 µm) avoids potential contamination during gas exchange
- Does not absorb cytokines and growth factors, and cell harvesting by conventional trypsin digestion method is possible
- Sterilised by irradiation
- Free from DNase and RNase, non pyrogenic
Documents
Avantor Cell Culture Solutions.pdf VWR for Cell Culture
Product Literature
CoC VWR Cell Chamber Tissue Culture Treated Vent Cap Sterile CoC_VWRI734-2980_210325530 CoC_VWRI734-2980_220514530A COC_VWRI734-2980_221009530F COC_VWRI734-2980_221109530F
More
CoC VWR Cell Chambers.pdf
Conformity declaration
TDS VWR® Cell Chamber.pdf
Datasheet
Specifications for Product Family
Specifications
- Catalog No:
- 734-2980
- 734-2980KR
- 734-2981
- 734-2981KR
- 734-2982
- 734-2982KR
- 734-2983
- 734-2983KR
- 734-3703
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- ISO 14001/Certificate No.: 04923E00243R0M
- ISO 14001/Certificate No.: 04923E00243R0M
- ISO 14001/Certificate No.: 04923E00243R0M
- ISO 14001/Certificate No.: 04923E00243R0M
Specifications
Product Family Options
Product Information
- TypeRecommended working volumeGrowth areaPackedAvailabilityPrice
- Specifications:
Low Manufacturing Impact More
Specifications
Cat. No.734-2980Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001/Certificate No.: 04923E00243R0MLow Manufacturing Impact - Specifications:
More
Specifications
Cat. No.734-2980KREnvironmentally PreferableEnvironmental certification - Specifications:
Low Manufacturing Impact More
Specifications
Cat. No.734-2981Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001/Certificate No.: 04923E00243R0MLow Manufacturing Impact - Specifications:
More
Specifications
Cat. No.734-2981KREnvironmentally PreferableEnvironmental certification - Specifications:
Low Manufacturing Impact More
Specifications
Cat. No.734-2982Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001/Certificate No.: 04923E00243R0MLow Manufacturing Impact - Specifications:
More
Specifications
Cat. No.734-2982KREnvironmentally PreferableEnvironmental certification - Specifications:
Low Manufacturing Impact More
Specifications
Cat. No.734-2983Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001/Certificate No.: 04923E00243R0MLow Manufacturing Impact - Specifications:
More
Specifications
Cat. No.734-2983KREnvironmentally PreferableEnvironmental certification - Specifications:New Product
More
Specifications
Cat. No.734-3703Environmentally PreferableEnvironmental certificationNew Product