Order Entry
Japan
Orders LinkContactUsLinkComponent
VWR®, Filter forceps
114 mm
Catalog # VWRU89259-954
Supplier:   VWR Collection
VWR®, Filter forceps
Catalog # VWRU89259-954
Supplier:  VWR Collection
VWR®, Filter forceps
VWR®, Filter forceps
114 mm
Price
$10.31
Log in for availability
1 * 1 items
Product Details & Documents
These stainless steel forceps with black PP non-slip finger grips are ideal for handling membranes.
Specifications
  • Length:
    114 mm