VWR®, Precision tweezers
Catalog # VWRU89259-984
Supplier: VWR Collection
Specifications
- Description:Precision tweezers, style no. 3
- Length:120 mm
- Pk:1
Specifications
About this item
These precision tweezers are made from stainless steel and feature very sharp, fine points.
- Anti-magnetic properties
- Resistant to acids