Stearic acid, powder, GenAR® NF vegetable grade 50, Macron Fine Chemicals™
2236-24
:
- Pack type:Fibre drum
- Pk:100 lb
- Seq:7167
- Meets N.F. Requirements:Passes Test
- Meets J.P. Chemical Specifications:194 - 212
- Meets E.P. Chemical Specifications:Passes Test
- GMP Manufactured Product:≤0.1%
- Meets B.P. Chemical Specifications:Passes Test
- Preserve in well-closed containers.:Passes Test
- CAUTION: For Manufacturing, processing or repackaging:≤4.0
- Bulk Pharmaceutical Chemical:53 - 59 °C
- NF - Identification A:Passes Test
- NF - Identification B (Acid Value):194 - 212
- NF - Identification C:Passes Test
- NF - Residue on Ignition:≤ 0.1 %
- NF - Acidity:Passes Test
- NF - Color of Solution:Passes Test
- NF - Iodine Value:≤ 4.0
- NF - Freezing Point:53 - 59 °C
- NF - Assay (C₁₈H₃₆O₂) (stearic acid):40.0 - 60.0 %
- NF - Assay (Total of Stearic & Palmitic):≥ 90.0 %
- EP/BP - Identification A:Passes Test
- EP/BP - Identification B:Passes Test
- EP/BP - Identification C:Passes Test
- EP/BP - Appearance:Passes Test
- EP/BP - Acidity:Passes Test
- EP/BP - Iodine Value:≤ 4.0
- EP/BP - Freezing Point:53 - 59 °C
- EP/BP - Stearic Acid:40.0 - 60.0 %
- EP/BP - Stearic and Palmitic Acids:≥ 90.0 %
- JP - Stearic and Palmitic Acids:≥ 90.0 %
- JP - Stearic Acid:40.0 - 60.0 %
- JP - Acid Value:194 - 212
- JP - Iodine Value:≤ 4.0
- JP - Acidity:Passes Test
- JP - Heavy Metals (as Pb):≤ 20 ppm
- JP - Congealing Point:53 - 59 °C
- JP - Residue on Ignition:≤ 0.10 %
- Thru U.S. No. 30 Sieve:99.5 - 100.0 %
- Thru U.S. No. 100 Sieve:95.0 - 100.0 %
:
:284.48 g/mol
:13,08882
:361 °C (1013 hPa)
:67 °C
:0.94 g/cm³ (20 °C)
:Management of Change (MOC) category = R
:196 °C (closed cup)
:Refrigerator
:200-313-4
:57-11-4
:C₁₈H₃₆O₂
:MFCD00002752