Stearic acid 92% NF, Macron Fine Chemicals™
Catalog # 2025-24
Supplier: MACRON AVANTOR BRAND
CAS Number:
Specifications
- Pack type:Fiber drum, Leverpack®
- Pk:100 lb
- Seq:7167
Specifications
Specification Test Results
- Bulk Pharmaceutical Chemical:Passes Test
- NF - Identification A:Passes Test
- NF - Identification B (Acid Value):194 - 212
- NF - Identification C:Passes Test
- NF - Stearic and Palmitic Acids:≥96.0 %
- NF - Stearic Acid:≥90.0 %
- NF - Freezing Point:64 - 69 °C
- NF - Iodine Value:≤1.5
- NF - Residue on Ignition:≤0.1 %
- NF - Acidity:Passes Test
- NF - Color of Solution:Passes Test
Specification Test Results
About this item
:
:284.48 g/mol
Melting Pt:64…69 °C
Density:0.94 g/cm³
CAS Number:57-11-4
Formula:C₁₈H₃₆O₂
MDL Number:MFCD00002752