Sodium molybdate(VI) dihydrate 99.5-103.0%, BAKER ANALYZED® ACS, J.T.Baker®
Catalog # 3764-01
Supplier: Avantor
CAS Number:
Specifications
- Pack type:Poly bottle, white, blue T/E cap
- Pk:500 g
- Seq:3645
Specifications
Specification Test Results
- Meets ACS Reagent Chemical Requirements:—
- For Laboratory,Research,or Manufacturing Use:—
- Appearance:Passes Test
- Assay (Na₂MoO₄·2H₂O):99.5 - 103.0 %
- Insoluble Matter:≤0.005 %
- pH of 5% Solution at 25°C:7.0 - 10.5
- Chloride (Cl):≤0.005 %
- Ammonium (NH₄):≤0.001 %
- Sulfate (SO₄):≤0.015 %
- Trace Impurities - Iron (Fe):≤0.001 %
- Phosphate (PO₄):≤5 ppm
- Trace Impurities - ACS - Heavy Metals (as Pb):≤5 ppm
Specification Test Results
About this item
:
:241.95 g/mol
Merck Index:13,08718
Melting Pt:687 °C
Density:3.28 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-551-7
CAS Number:10102-40-6
Formula:Na₂MoO₄·2H₂O
MDL Number:MFCD00149170