

Methyl ethyl ketone ≥99.0% (by GC, corrected for water content), AR® ACS, Macron Fine Chemicals™
Catalog # 6240-28
Supplier: MACRON AVANTOR BRAND
CAS Number:
Specifications
- Pack type:Steel drum, phenolic lined
- Pk:200 L
- Seq:5549
Specifications
Specification Test Results
- Meets ACS Reagent Chemical Requirements:—
- For Laboratory,Research,or Manufacturing Use:≥99.0%
- ACS - Assay (CH₃COCH₂CH₃) (by GC, corrected for water):≥99.0 %
- ACS - Color (APHA):≤15
- ACS - Residue after Evaporation:≤0.0025 %
- ACS - Titrable Acid (meq/g):≤0.0005
- ACS - Water (by KF, volumetric):≤0.20 %
Specification Test Results
About this item
:
Merck Index:13,06097
Boiling Pt:79.6 °C (1013 hPa)
Melting Pt:–86 °C
Density:0.8054 g/cm³ (20 °C)
Flash Pt:–4 °C
ADR:3,II
:Management of Change (MOC) category = N
:72.11 g/mol
Storage Temperature:Ambient
EINECS:201-159-0
UN:1193
REACH:01-2119457290-43
:GHS02
:GHS07
CAS Number:78-93-3
Formula:C₂H₅COCH₃
MDL Number:MFCD00011648