Order Entry
Japan
ContactUsLinkComponent
 
 
Guanidinium thiocyanate ≥99.0% (dried basis), BAKER ANALYZED®, Biochemical Reagent, J.T.Baker®
  4083-01
 :  Avantor
 :  
undefined
 
Guanidinium thiocyanate ≥99.0% (dried basis), BAKER ANALYZED®, Biochemical Reagent, J.T.Baker®
  4083-01
 :  Avantor
 :  

 

  • Pack type:
    Poly bottle
  • Pk:
    500 g
  • Seq:
    3645

 

 

  • Assay (NH₂C(:NH)NH₂·HSCN) (dried basis):
    ≥99.0 %
  • Identification:
    Passes Test
  • Loss on Drying at 105°C:
    ≤0.5 %
  • pH of 5% Solution at 25°C:
    4.0 - 7.0
  • Absorbance of a 1.7M Solution - 340 nm:
    ≤0.030
  • Absorbance of a 1.7M Solution - 300 nm:
    ≤0.050
  • Absorbance of a 1.7M Solution - 280 nm:
    ≤0.300
  • DNase Activity (None Detected):
    Passes Test
  • Protease Activity (None Detected):
    Passes Test
  • RNase Activity (None Detected):
    Passes Test
  • Trace Impurities - Arsenic (As):
    ≤1 ppm
  • Trace Impurities - Copper (Cu):
    ≤1 ppm
  • Trace Impurities - Iron (Fe):
    ≤1 ppm
  • Trace Impurities - Lead (Pb):
    ≤2.000 ppm

 

 

 :
 :118.16 g/mol
 :118 °C
 :1.29 g/cm³ (20 °C)
 :209-812-1
 :GHS07
 :Ambient
 :593-84-0
 :CH₅N₃·CHNS
 :MFCD00013027