
Calcium chloride, anhydrous 93.0-100.5%, CALTAC FCC, Macron Fine Chemicals™
Catalog # 4880-32
Supplier: MACRON AVANTOR BRAND
CAS Number:
Specifications
- Pack type:Bag
- Pk:50 lb
- Seq:7137
Specifications
Specification Test Results
- Food GMP Manufactured Product:93.0 - 100.5%
- Meets F.C.C. Requirements:Passes Test
- Preserve in Tight Containers:Passes Test
- CAUTION: For Manufacturing, processing or repackaging:≤0.02%
- Bulk Food Chemical:Passes Test
- FCC - Assay (CaCl₂):93.0 - 100.5 %
- FCC - Identification A:Passes Test
- FCC - Identification B:Passes Test
- Solubility:Passes Test
- Acid-Insoluble Substances - Retained Mass:≤0.02 %
- Acid-Insoluble Substances - No Particles >2mm:Passes Test
- Arsenic (As):≤3 mg/kg
- Fluoride (F):≤0.004 %
- FCC - Lead (Pb):≤5 mg/kg
- Magnesium and Alkali Salts:≤5.0 %
- pH (1 in 20):4.5 - 11.0
Specification Test Results
About this item
:
Merck Index:14,01659
Boiling Pt:1600 °C (1013 hPa)
Melting Pt:772 °C
Density:2.15 g/cm³ (20 °C)
:110.98 g/mol
Storage Temperature:Ambient
EINECS:233-140-8
REACH:01-2119494219-28
:GHS07
CAS Number:10043-52-4
Formula:CaCl₂
MDL Number:MFCD00010903