Order Entry
Japan
ContactUsLinkComponent
Bismuth(III) oxide ≥99.7%, AR®, Macron Fine Chemicals™
Bismuth(III) oxide ≥99.7%, AR®, Macron Fine Chemicals™
Catalog # 1057-24
CAS Number:  
undefined
Bismuth(III) oxide ≥99.7%, AR®, Macron Fine Chemicals™
Catalog # 1057-24
CAS Number:  

Specifications

  • Pack type:
    Fibre drum
  • Pk:
    100 lb
  • Seq:
    7167

Specifications

Specification Test Results

  • CAUTION: For Manufacturing, processing or repackaging:
    ≥99.7%
  • Assay (Bi₂O₃):
    ≥ 99.7 %
  • pH (by aqueous extraction):
    6.75 - 9.25
  • Appearance (yellow powder):
    Passes Test
  • Arsenic (As):
    ≤ 0.0008 %
  • Chloride (Cl):
    ≤ 0.001 %
  • Copper (Cu):
    ≤ 0.01 %
  • Iron (Fe):
    ≤ 0.01 %
  • Sodium (Na):
    ≤ 0.03 %
  • On U.S. No. 325 Sieve:
    ≤ 1.0 %

Specification Test Results

About this item

 :
 :465.96 g/mol
Density:8.90 g/cm³
 :Management of Change (MOC) category = RL
CAS Number:1304-76-3
Formula:BiO₃
MDL Number:MFCD00003462