Order Entry
Japan
ContactUsLinkComponent
Zinc phenolsulphonate 73.7 - 77.4 %, granular, purified, Macron Fine Chemicals™
Irritant
Zinc phenolsulphonate 73.7 - 77.4 %, granular, purified, Macron Fine Chemicals™
Certificates
undefined
Irritant
Zinc phenolsulphonate 73.7 - 77.4 %, granular, purified, Macron Fine Chemicals™

Specification Test Results

  • Assay (Zn(C₆H₅O₄S)₂):
    73.7 - 77.4 %
  • Appearance:
    Passes Test
  • Identification A:
    Passes Test
  • Identification B:
    Passes Test
  • Identification C:
    Passes Test
  • Sulfate (SO₄):
    ≤0.04 %
  • Solubility (clear, colorless solution):
    Passes Test
  • Arsenic (As):
    ≤0.0003 %
  • Lead (Pb):
    ≤0.0020 %
  • pH of 0.4% Solution at 25°C, For Information Only:
    ≤10
  • Iron (Fe):
    ≤0.0005 %
  • Solubility in Alcohol (NTU):
    ≤10

Specification Test Results

Frequently Bought Together