Order Entry
Japan
ContactUsLinkComponent
VWR® Beakers, Disposable
  213-3918
 :
undefined
VWR® Beakers, Disposable
  213-3918
 :
Select an Option
64 mm
Select an Option
5 - 100 ml
Select an Option
100 ml
Select an Option
62 mm
Select an Option
04
Select an Option
100
PP, translucent.
  • Embossed graduations
  • Ribs for easy stacking
  • Autoclavable to 121 °C
  • With spout
High quality economical beaker for everyday work. Ideal for general laboratory use.
 : *The product(s) on this page are outside the scope of the EU Directive 2019/904
Frequently Bought Together