Order Entry
Japan
Orders LinkContactUsLinkComponent
VWR®, Fittings for Capillary Connector
VWR®, Fittings for Capillary Connector
Supplier:   VWR Collection
VWR®, Fittings for Capillary Connector
Supplier:  VWR Collection
Certificates
VWR®, Fittings for Capillary Connector
VWR®, Fittings for Capillary Connector
4 Options Available Below
Product Details & Documents
Choose the suitable fitting for your tubing size.
Documents
Product Family Options
Product Information
  • Material
    Colour
    Availability
    Price
  • 590-0042
    Fitting with integrated ferrule, 1,6 mm OD
    Fitting with integrated ferrule, 1,6 mm OD
    PFA
    Green
    Log in for availability
    Log in for availability
    1 * 5 items
    1 * 5 items
  • 590-0041
    Fitting with integrated ferrule, 2,3 mm OD
    Fitting with integrated ferrule, 2,3 mm OD
    PFA
    Grey
    Log in for availability
    Log in for availability
    1 * 5 items
    1 * 5 items
  • 590-0040
    Fitting with integrated ferrule, 3,2 mm OD
    Fitting with integrated ferrule, 3,2 mm OD
    PFA
    Black
    Log in for availability
    Log in for availability
    1 * 5 items
    1 * 5 items
  • 590-0039
    Blind plug for capillary connector
    Blind plug for capillary connector
    PFA
    Colourless
    Log in for availability
    Log in for availability
    1 * 10 items
    1 * 10 items