Order Entry
Japan
ContactUsLinkComponent
VWR®, Jerrican, Narrow Mouth, HDPE, Rectangular
VWR®, Jerrican, Narrow Mouth, HDPE, Rectangular
Catalog # 216-1917
Supplier:  VWR Collection
undefined
VWR®, Jerrican, Narrow Mouth, HDPE, Rectangular
Catalog # 216-1917
Supplier:  VWR Collection

Specifications

  • Capacity:
    20 l
  • Seq:
    01
  • Neck I-Ø:
    53 mm
  • Pk:
    1

Specifications

About this item

Disposable jerrican, made of HDPE, with integrated handle for easy carrying and pouring.

  • Good chemical resistance
  • Leakproof on standard test conditions
  • Graduated to identify the volume dispensed
  • Angled neck opening for easy pouring and transferring liquid

Jerricans are ideal for storage and transfer of sterile fluids, as well as pharmaceutical and biotechnology reagents.

Certifications: Resin meets US FDA 21 CFR, USP Class VI.